Anti-UBTF Rabbit Monoclonal Antibody [clone: 13A226]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029636EA 3560 DKK
USBI029636
Anti-UBTF Rabbit Monoclonal Antibody [clone: 13A226]
Antistoffer
Recommended dilution: Flow Cytometry: 1:30; Western Blot: 1:1000-10000; Immunochemistry: 1:100-1:250;

Type: Primary
Antigen: UBTF (upstream binding transcription factor, RNA polymerase I)
Clonality: Monoclonal
Clone: 13A226
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR