Anti-TERT Mouse Monoclonal Antibody [clone: 13A223]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029623EA 4150 DKK
USBI029623
Anti-TERT Mouse Monoclonal Antibody [clone: 13A223]
Antistoffer
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: Monoclonal
Clone: 13A223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR