Anti-Tensin-3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029622EA 4220 DKK
USBI029622
Anti-Tensin-3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: Tensin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR