Anti-TAF6 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029619EA 3310 DKK
USBI029619
Anti-TAF6 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: TAF6 (TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR