Anti-SMARCA4 Mouse Monoclonal Antibody [clone: 13A219]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029611EA 4290 DKK
USBI029611
Anti-SMARCA4 Mouse Monoclonal Antibody [clone: 13A219]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: SMARCA4 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4)
Clonality: Monoclonal
Clone: 13A219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR