Anti-PRDM12 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029587EA 4290 DKK
USBI029587
Anti-PRDM12 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 3?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: PRDM12 (PR domain containing 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR