Anti-RBFOX3 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029556EA 4870 DKK
USBI029556
Anti-RBFOX3 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: RBFOX3 (RNA binding protein, fox-1 homolog (C. elegans) 3)
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR