Anti-MAP2 Mouse Monoclonal Antibody (Cy3®) [clone: 13A207]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029534EA 4850 DKK
USBI029534
Anti-MAP2 Mouse Monoclonal Antibody (Cy3®) [clone: 13A207]
Antistoffer
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Monoclonal
Clone: 13A207
Conjugation: Cy3®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR