Anti-MAP2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029533EA 5190 DKK
USBI029533
Anti-MAP2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR