Anti-DFFA Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029510EA 4110 DKK
USBI029510
Anti-DFFA Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:500

Type: Primary
Antigen: DFFA (DNA fragmentation factor, 45kDa, alpha polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR