Anti-Histone H3, Monomethyl, Dimethyl, Trimethyl Rabbit Monoclonal Antibody [clone: 13A193]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029498EA 5230 DKK
USBI029498
Anti-Histone H3, Monomethyl, Dimethyl, Trimethyl Rabbit Monoclonal Antibody [clone: 13A193]
Antistoffer
Recommended dilution: Western Blot: 1:8000

Type: Primary
Antigen: Histone H3, Monomethyl, Dimethyl, Trimethyl
Clonality: Monoclonal
Clone: 13A193
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR