Anti-HELLS Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029494EA 4190 DKK
USBI029494
Anti-HELLS Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 2?g/ml; Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: HELLS (helicase, lymphoid-specific)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR