Anti-H2AFX Rabbit Monoclonal Antibody [clone: 13A191]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029491EA 4250 DKK
USBI029491
Anti-H2AFX Rabbit Monoclonal Antibody [clone: 13A191]
Antistoffer
Recommended dilution: Western Blot: 1:1000-10000; Immunochemistry: 1:50-1:100;-1:100

Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: Monoclonal
Clone: 13A191
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR