Anti-GFAP Mouse Monoclonal Antibody (Biotin) [clone: 13A187]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI029483EA 5850 DKK
USBI029483
Anti-GFAP Mouse Monoclonal Antibody (Biotin) [clone: 13A187]
Antistoffer
Recommended dilution: Immunochemistry: 1:100
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: Monoclonal
Clone: 13A187
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human