Anti-Galactocerebroside Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A186]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029482EA 3730 DKK
USBI029482
Anti-Galactocerebroside Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A186]
Antistoffer
Type: Primary
Antigen: Galactocerebroside
Clonality: Monoclonal
Clone: 13A186
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR