Anti-FOXO1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029480EA 3780 DKK
USBI029480
Anti-FOXO1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: FOXO1 (forkhead box O1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR