Anti-FABP4 Rabbit Monoclonal Antibody [clone: 13A183]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029470EA 3740 DKK
USBI029470
Anti-FABP4 Rabbit Monoclonal Antibody [clone: 13A183]
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:5000

Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: Monoclonal
Clone: 13A183
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR