Anti-DNP Mouse Monoclonal Antibody [clone: 13A173]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029454EA 4380 DKK
USBI029454
Anti-DNP Mouse Monoclonal Antibody [clone: 13A173]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: Monoclonal
Clone: 13A173
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR