Anti-CACNA1D Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029431EA 4110 DKK
USBI029431
Anti-CACNA1D Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.1 ?g/ml; Immunochemistry: 1:200

Type: Primary
Antigen: CACNA1D (calcium channel, voltage-dependent, L type, alpha 1D subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR