Anti-BMPR1A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029427EA 4290 DKK
USBI029427
Anti-BMPR1A Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: BMPR1A (bone morphogenetic protein receptor, type IA)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR