Anti-BMP4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A161]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029426EA 3650 DKK
USBI029426
Anti-BMP4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A161]
Antistoffer
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: Monoclonal
Clone: 13A161
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR