Anti-ATG4D Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029422EA 4620 DKK
USBI029422
Anti-ATG4D Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:200

Type: Primary
Antigen: ATG4D (autophagy related 4D, cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR