Anti-SOD3 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029372EA 2990 DKK
USBI029372
Anti-SOD3 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: SOD3 (superoxide dismutase 3, extracellular)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR