Anti-RuBisCO Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029349EA 2770 DKK
USBI029349
Anti-RuBisCO Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: RuBisCO
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR