Anti-RPL8 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029301EA 3180 DKK
USBI029301
Anti-RPL8 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: RPL8 (ribosomal protein L8)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR