Anti-RAB1A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029284EA 3180 DKK
USBI029284
Anti-RAB1A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RAB1A (RAB1A, member RAS oncogene family)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR