Anti-PFDN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029239EA 2990 DKK
USBI029239
Anti-PFDN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: PFDN1 (prefoldin subunit 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR