Anti-CSTB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029102EA 3180 DKK
USBI029102
Anti-CSTB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: CSTB (cystatin B (stefin B))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR