Anti-CSNK1E Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029097EA 3180 DKK
USBI029097
Anti-CSNK1E Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CSNK1E (casein kinase 1, epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR