Anti-COX6A1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029092EA 3180 DKK
USBI029092
Anti-COX6A1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COX6A1 (cytochrome c oxidase subunit VIa polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR