Anti-ATP5D Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029052EA 2990 DKK
USBI029052
Anti-ATP5D Rabbit Polyclonal Antibody
Type: Primary
Antigen: ATP5D (ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR