Anti-CD40L Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MR1]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-B08-N91EA 3710 DKK
ROCK200-B08-N91
Anti-CD40L Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MR1]
Antistoffer
Anti-CD154 is useful for immunochemistry and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: MR1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR