Anti-AKT2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11F6.B2]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-526-J35EA 2890 DKK
ROCK200-526-J35
Anti-AKT2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11F6.B2]
Antistoffer
Anti-AKT2 APC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT2 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 11F6.B2
Conjugation: APC (Allophycocyanin)
Epitope: Center
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR