Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 3C7]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-508-N82EA 6070 DKK
ROCK200-508-N82
Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 3C7]
Antistoffer
Anti-CD25 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 3C7
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR