Anti-CD45.1 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: A20]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-502-N85EA 2600 DKK
ROCK200-502-N85
Anti-CD45.1 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: A20]
Antistoffer
Anti-CD45.1 is useful for Flow Cytometry, Immunoprecipitation, and immunochemistry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR