Anti-AGAP4 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-401-P07EA 5840 DKK
ROCK200-401-P07
Anti-AGAP4 Rabbit Polyclonal Antibody
Antistoffer
Anti-AGAP4 antibody has been tested for use in western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 73kDa in size corresponding to AGAP4 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000

Type: Primary
Antigen: AGAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR