Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-326-N70EA 7120 DKK
ROCK200-326-N70
Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]
Antistoffer
Anti-CD45RA is useful for Flow Cytometry, immunochemistry, and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: HI100
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR