Anti-AKT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 14E5.A2.B2.H9]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-326-I51EA 2890 DKK
ROCK200-326-I51
Anti-AKT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 14E5.A2.B2.H9]
Antistoffer
Anti-AKT1 APC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: Flow Cytometry: 1.0 µg/ml;
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: APC (Allophycocyanin)
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR