Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 5E5.F5.D7]
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROCK200-306-J34EA 3780 DKK
ROCK200-306-J34
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 5E5.F5.D7]
Antistoffer
Anti-AKT3 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 5E5.F5.D7
Conjugation: Biotin
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human