Anti-AKT3 Mouse Monoclonal Antibody (Biotin) [clone: 25F6.F6.D8]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-306-E75EA 2890 DKK
ROCK200-306-E75
Anti-AKT3 Mouse Monoclonal Antibody (Biotin) [clone: 25F6.F6.D8]
Antistoffer
Anti-AKT3 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 25F6.F6.D8
Conjugation: Biotin
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR