Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 5C3]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-302-N67EA 2600 DKK
ROCK200-302-N67
Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 5C3]
Antistoffer
Anti-CD40 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells
Do not freeze!

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR