Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 4F2.D9.C5.G5.B2.C5]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK200-301-I49SEA 795 DKK
ROCK200-301-I49S
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 4F2.D9.C5.G5.B2.C5]
Antistoffer
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 4F2.D9.C5.G5.B2.C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR