Anti-IgG Mouse Monoclonal Antibody (DyLight® 680)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK18-4416-32EA 1700 DKK
ROCK18-4416-32
Anti-IgG Mouse Monoclonal Antibody (DyLight® 680)
Antistoffer
Fluorescent Rabbit TrueBlot® Antibody DyLight® 680 is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB182
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR