Anti-MORC3 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK100-401-N97EA 2830 DKK
ROCK100-401-N97
Anti-MORC3 Rabbit Polyclonal Antibody
Antistoffer
Anti-Morc3 Antibody is suitable for use in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 107.1 kDa in size corresponding Morc3 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:20000-1:50000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: MORC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR