Anti-TET3 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK100-401-N94EA 5840 DKK
ROCK100-401-N94
Anti-TET3 Rabbit Polyclonal Antibody
Antistoffer
Tet3 antibody has been tested for use by ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect band at ~180kDa.

Recommended Dilutions: ELISA: 1:50000; Western Blot: 1:5000;

Type: Primary
Antigen: TET3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR