IgG1 Isotype Control (APC (Allophycocyanin))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK010-2640EA 3570 DKK
ROCK010-2640
IgG1 Isotype Control (APC (Allophycocyanin))
Antistoffer
Mouse IgG1 isotype control can be utilized as a control or standard reagent in Flow Cytometry, Western Blotting, and ELISA experiments where determination of sample isotype is important.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR