Anti-IgG (H+L) Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK807-1102EA 1740 DKK
ROCK807-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Golden Syrian Hamster
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR