Anti-IgG (H+L) Goat Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK712-1833EA 2100 DKK
ROCK712-1833
Anti-IgG (H+L) Goat Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: Immunofluorescence: 1:100 - 1:250;
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR