Anti-IgG (H+L) Donkey Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK710-701-124EA 1100 DKK
ROCK710-701-124
Anti-IgG (H+L) Donkey Polyclonal Antibody
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR