Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK710-1932EA 1570 DKK
ROCK710-1932
Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)
Antistoffer
Secondary Goat Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR